Invitrogen
Manufacturer Code:PA5110208
Catalog # PIPA5110208
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human PPP2R1A (NP_055040.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 29B 6330556D22Rik A/PR65 BOS_17968 CG13383 CG17291 CG33297 DmelCG17291 Dmel_CG17291 dPP2A A DPR65 Medium tumor antigen-associated 61 kDa protein MGC786 MRD36 Pp2A PP2A 29B PP2A A PP2A subunit A isoform PR65-alpha PP2A subunit A isoform R1-alpha PP2A[A] Pp2A29B Pp2A-29B PP2A-A PP2AAALPHA PP2A-Aalpha PP2Ac PP2CA PP2Calpha PPP2R1A PR65 PR65A Protein phosphatase PP2A regulatory subunit A RP-C Serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alpha isoform Serine/threonine-protein phosphatase PP2A 65 kDa regulatory subunit; Medium tumor antigen-associated 61 kDa protein; PP2A subunit A isoform PR65-alpha; PP2A subunit A isoform R1-alpha; protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), alpha isoform; protein phosphatase 2 regulatory subunit A, alpha; protein phosphatase 2, regulatory subunit A, alpha; protein phosphatase PP2A; serine/threonine protein phosphatase 2A, 65 kDa regulatory subunit A, alpha isoform; serine/threonine protein phosphatase A subunit type 2A; Serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alpha isoform; testicular secretory protein Li 1
Gene Aliases: 6330556D22Rik; MRD36; PP2A; PP2A-Aalpha; PP2AAALPHA; PPP2R1A; PR65; PR65A
UniProt ID: (Human) P30153, (Mouse) Q76MZ3
Entrez Gene ID: (Human) 5518, (Mouse) 51792
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.