Abnova
Manufacturer Code:H00005518K
Catalog # 89-210-037
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human PPP2R1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human PPP2R1A peptide using ARM Technology.
For Research Use Only
Protein Aliases: Medium tumor antigen-associated 61 kDa protein; MGC786 PR65A; PP2A subunit A isoform PR65-alpha; PP2A subunit A isoform R1-alpha; protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), alpha isoform; protein phosphatase 2 regulatory subunit A, alpha; protein phosphatase 2, regulatory subunit A, alpha; serine/threonine protein phosphatase 2A, 65 kDa regulatory subunit A, alpha isoform; Serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alpha isoform; testicular secretory protein Li 1
Gene Aliases: MRD36; PP2A-Aalpha; PP2AAALPHA; PPP2R1A; PR65A
UniProt ID: (Human) P30153
Entrez Gene ID: (Human) 5518
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.