Invitrogen
Manufacturer Code:PA5111734
Catalog # PIPA5111734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human PPP1R9A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810430P21Rik 4930518N04Rik 5330407E15 A230094E16Rik BB181831 KIAA1222 mKIAA1222 Neb1 Neurabin-1 Neurabin-I Neural tissue-specific F-actin-binding protein I NRB NRB1 NRBI p180 PP1bp175 PPP1R9A Protein phosphatase 1 regulatory subunit 9A; Neurabin-1; Neurabin-I; Neural tissue-specific F-actin-binding protein I; Protein phosphatase 1 regulatory subunit 9A; protein phosphatase 1, regulatory (inhibitor) subunit 9A; protein phosphatase 1, regulatory subunit 9A
Gene Aliases: KIAA1222; Neurabin-I; NRB1; NRBI; PPP1R9A
UniProt ID: (Human) Q9ULJ8
Entrez Gene ID: (Human) 55607
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.