Proteintech
Manufacturer Code:188011AP150UL
Catalog # 50-173-2229
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: actin-binding protein neurabin; KIAA1222 Neurabin 1 Neurabin I NRB1 NRBI PPP1R9A; neurabin 1; Neurabin I; Neurabin-1; Neurabin-I; neural tissue-specific F-actin binding protein; Neural tissue-specific F-actin-binding protein I; p180; PP1bp175; Protein phosphatase 1 regulatory subunit 9A; protein phosphatase 1, regulatory (inhibitor) subunit 9A; protein phosphatase 1, regulatory subunit 9A
Gene Aliases: 2810430P21Rik; 4930518N04Rik; 5330407E15; A230094E16Rik; BB181831; KIAA1222; mKIAA1222; Neb1; Neurabin-I; NRB; NRB1; NRBI; PPP1R9A
UniProt ID: (Human) Q9ULJ8, (Rat) O35867
Entrez Gene ID: (Human) 55607, (Mouse) 243725, (Rat) 84685
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.