Invitrogen
Manufacturer Code:PA555601
Catalog # PIPA555601
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PPP1R8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activator of RNA decay; ARD-1; NIPP-1; Nuclear inhibitor of protein phosphatase 1; nuclear inhibitor of protein phosphatase-1 alpha; nuclear inhibitor of protein phosphatase-1 beta; nuclear subunit of PP-1; PPP1R8 NIPP1 protein phosphatase 1 regulatory (inhibitor) subunit 8 ARD1 NIPP-1 PRO2047 ard-1 activator of RNA decay nuclear subunit of PP-1 homolog of E.coli RNase E nuclear inhibitor of protein phosphatase-1 protein phosphatase-1 regulatory subunit-8 ARD-1 RNase E protein phosphatase 1 regulatory inhibitor subunit 8 protein phosphatase 1 regulatory subunit 8; Protein phosphatase 1 regulatory inhibitor subunit 8; protein phosphatase 1, regulatory (inhibitor) subunit 8; protein phosphatase 1, regulatory subunit 8; RNase E
Gene Aliases: 6330548N22Rik; ARD-1; ARD1; AU044684; NIPP-1; NIPP1; PPP1R8; PRO2047
UniProt ID: (Human) Q12972, (Mouse) Q8R3G1
Entrez Gene ID: (Human) 5511, (Rat) 313030, (Mouse) 100336
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.