Novus Biologicals
Manufacturer Code:NBP234170
Catalog # NBP234170
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: DAKKEPGAECFHFSLCLPPGLQPEDEEDADERGVAVHFAVCYRCAQGEYWDNNAGANYTLRY |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein phosphatase 1 regulatory subunit 3G; Protein Phosphatase 1 Regulatory Subunit 3G Protein Phosphatase 1 Regulatory (Inhibitor) Subunit 3G Protein Phosphatase 1 Regulatory Subunit 3G Putative Protein Phosphatase 1 Regulatory Inhibitor Subunit 3G; protein phosphatase 1, regulatory (inhibitor) subunit 3G; protein phosphatase 1, regulatory subunit 3G; putative protein phosphatase 1 regulatory inhibitor subunit 3G
Gene Aliases: PPP1R3G
UniProt ID: (Human) B7ZBB8
Entrez Gene ID: (Human) 648791
Molecular Function: enzyme modulator phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.