Invitrogen
Manufacturer Code:PA5113067
Catalog # PIPA5113067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Protein phosphatase 1 regulatory subunit 3B protein (1-285AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 33 kDa glycogen-binding protein; 6430576E21 AW821953 GL Hepatic glycogen-targeting protein phosphatase 1 regulatory subunit GL hepatic glycogen-targeting subunit G(L) PP1 subunit R4 PPP1R3B PPP1R4 Protein phosphatase 1 regulatory subunit 3B Protein phosphatase 1 regulatory subunit 4 Protein phosphatase 1 subunit GL protein phosphatase 1 regulatory (inhibitor) subunit 3B protein phosphatase 1 regulatory subunit 3B PTG; Hepatic glycogen-targeting protein phosphatase 1 regulatory subunit GL; hepatic glycogen-targeting subunit, G(L); PP1 subunit R4; protein phosphatase 1 (GL-subunit); Protein phosphatase 1 regulatory subunit 3B; Protein phosphatase 1 regulatory subunit 4; Protein phosphatase 1 subunit GL; protein phosphatase 1, regulatory (inhibitor) subunit 3B; protein phosphatase 1, regulatory subunit 3B; PTG; R4
Gene Aliases: 6430576E21; AW821953; GL; PPP1R3B; PPP1R4; PTG
UniProt ID: (Human) Q86XI6, (Mouse) Q8C767, (Rat) Q6IN01
Entrez Gene ID: (Human) 79660, (Mouse) 244416, (Rat) 192280
Molecular Function:
enzyme modulator
phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.