Invitrogen
Manufacturer Code:PA5107219
Catalog # PIPA5107219
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide located within the following region: LSSKNEASGM VAPAVQEKKV KKRVSFADNQ GLALTMVKVF SEFDDPLDMP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hepatic glycogen-targeting protein phosphatase 1 regulatory subunit GL; hepatic glycogen-targeting subunit, G(L); PP1 subunit R4; Protein phosphatase 1 regulatory subunit 3B; Protein phosphatase 1 regulatory subunit 4; Protein phosphatase 1 subunit GL; protein phosphatase 1, regulatory (inhibitor) subunit 3B; protein phosphatase 1, regulatory subunit 3B; PTG
Gene Aliases: GL; PPP1R3B; PPP1R4; PTG
UniProt ID: (Human) Q86XI6
Entrez Gene ID: (Human) 79660
Molecular Function:
enzyme modulator
phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.