Abnova
Manufacturer Code:PAB9626
Catalog # 89-134-282
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to N-terminus rat Ppp1r1b. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Ppp1r1b.
For Research Use Only
Protein Aliases: DARPP-32; Darpp32; Dopamine- and cAMP-regulated neuronal phosphoprotein; dopamine- and cAMP-regulated phosphoprotein DARPP-32; neuronal phosphoprotein DARPP-32; Protein phosphatase 1 regulatory subunit 1B
Gene Aliases: Darpp-32; Darpp32; Ppp1r1b
UniProt ID: (Rat) Q6J4I0
Entrez Gene ID: (Rat) 360616
Molecular Function: enzyme modulator phosphatase inhibitor phosphatase modulator signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.