Abnova
Manufacturer Code:PAB25435
Catalog # 89-127-052
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding T75 of human PPP1R1B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PPP1R1B.
For Research Use Only
Protein Aliases: DARPP-32; DARPP32; dopamine and cAMP-regulated neuronal phosphoprotein 32; Dopamine- and cAMP-regulated neuronal phosphoprotein; FLJ20940; Protein phosphatase 1 regulatory subunit 1B
Gene Aliases: DARPP-32; DARPP32; PPP1R1B
UniProt ID: (Human) Q9UD71
Entrez Gene ID: (Human) 84152
Molecular Function: enzyme modulator phosphatase inhibitor phosphatase modulator signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.