Novus Biologicals
Manufacturer Code:NBP286752
Catalog # NB034374
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of PPP1R16B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANKRD4TIMAPankyrin repeat domain protein 4 Ankyrin repeat domain-containing protein 4 CAAX box protein TIMAP hTIMAP KIAA0823TGF-beta-inhibited membrane-associated protein protein phosphatase 1 regulatory inhibitor subunit 16B protein phosphatase 1 regulatory (inhibitor) subunit 16B; ankyrin repeat domain protein 4; Ankyrin repeat domain-containing protein 4; CAAX box protein TIMAP; hTIMAP; Protein phosphatase 1 regulatory inhibitor subunit 16B; protein phosphatase 1, regulatory (inhibitor) subunit 16B; protein phosphatase 1, regulatory subunit 16B; TGF-beta-inhibited membrane-associated protein
Gene Aliases: ANKRD4; KIAA0823; PPP1R16B; TIMAP
UniProt ID: (Human) Q96T49
Entrez Gene ID: (Human) 26051
Molecular Function: enzyme modulator phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.