Proteintech
Manufacturer Code:184761AP150UL
Catalog # 50-173-2163
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PPP1R14B Fusion Protein Ag13373 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PHI 1 PLCB3N PNG PPP1R14B SOM172; PHI-1; Phosphatase holoenzyme inhibitor 1; phospholipase C beta-3 neighboring gene protein; phospholipase c neighboring; Phospholipase C-beta-3 neighbouring gene protein; Protein phosphatase 1 regulatory subunit 14B; protein phosphatase 1, regulatory (inhibitor) 5 subunit 14B; protein phosphatase 1, regulatory (inhibitor) subunit 14B; Ubiquitous PKC-potentiated PP1 inhibitor
Gene Aliases: AOM172; PHI-1; Phi1; PLCB3N; PNG; PPP1R14B; SOM172
UniProt ID: (Human) Q96C90, (Rat) Q8K3F3, (Mouse) Q62084
Entrez Gene ID: (Human) 26472, (Rat) 259225, (Mouse) 18938
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.