Invitrogen
Manufacturer Code:PA548110
Catalog # PIPA548110
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit human PNUTS Pro5-Asn226 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HLA-C adjacent transcript 53; HLA-C associated transcript 53; MHC class I region proline-rich protein CAT53; p99; Phosphatase 1 nuclear targeting subunit; PP1-binding protein of 114 kDa; PPP1R10 CAT53 FB19PP1-binding protein of 114 kDa MHC class I region proline-rich protein CAT53 p99 Phosphatase 1 nuclear targeting subunit phosphatase nuclear targeting subunit PNUTSPP1R10 Protein FB19 protein phosphatase 1 regulatory subunit 10 protein phosphatase 1 regulatory (inhibitor) subunit 10 protein phosphatase 1 regulatory subunit 10 serine/threonine-protein phosphatase 1 regulatory subunit 10; Protein FB19; protein phosphatase 1, regulatory (inhibitor) subunit 10; protein phosphatase 1, regulatory subunit 10; protein phosphatase I; Protein PNUTS; putative protein phosphatase 1 nuclear targeting subunit; Serine/threonine-protein phosphatase 1 regulatory subunit 10; testis tissue sperm-binding protein Li 67n
Gene Aliases: 2610025H06Rik; CAT53; D17Ertd808e; FB19; p99; PNUTS; PP1R10; PPP1R10; R111
UniProt ID: (Human) Q96QC0, (Rat) O55000, (Mouse) Q80W00
Entrez Gene ID: (Human) 5514, (Rat) 65045, (Mouse) 52040
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.