Novus Biologicals
Manufacturer Code:NBP273583APC
Catalog # NB273583APC
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7F12 |
Immunogen | Full length recombinant protein of human PPME1 (NP_057231) produced in HEK293T cells. |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 3.1.1 EC 3.1.1.- FLJ22226 PME-1PME1 protein phosphatase methylesterase 1 protein phosphatase methylesterase-1; PME-1; Protein phosphatase methylesterase 1; protein phosphatase methylesterase-1; testicular secretory protein Li 39
Gene Aliases: PME-1; PME1; PP2593; PPME1; PRO0750
UniProt ID: (Human) Q9Y570
Entrez Gene ID: (Human) 51400
Molecular Function:
esterase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.