Abnova
Manufacturer Code:H00005496K
Catalog # 89-200-831
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human PPM1G |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human PPM1G peptide using ARM Technology.
For Research Use Only
Protein Aliases: MGC1675 MGC2870 PP2CG PP2CGAMMA PPP2CG; PP2C, gamma; PP2C-gamma; Protein phosphatase 1C; Protein phosphatase 1G; protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform; protein phosphatase 2, catalytic subunit, gamma isoform; protein phosphatase 2C gamma isoform; Protein phosphatase 2C isoform gamma; Protein phosphatase magnesium-dependent 1 gamma; protein phosphatase, Mg2+/Mn2+ dependent, 1G
Gene Aliases: PP2CG; PP2CGAMMA; PPM1C; PPM1G; PPP2CG
UniProt ID: (Human) O15355
Entrez Gene ID: (Human) 5496
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.