OriGene
Manufacturer Code:TA502865
Catalog # 50-167-3873
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3F2 |
Immunogen | Full length human recombit protein of human PPM1B produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: PP2C-beta; PP2C-beta PP2C-beta-X PP2CB PP2CBETA PPC2BETAX; Protein phosphatase 1B; protein phosphatase 1B (formerly 2C), magnesium-dependent, beta isoform; protein phosphatase 1B, magnesium dependent, beta isoform; Protein phosphatase 2C isoform beta; protein phosphatase 2C-like protein; Protein phosphatase type 1B (formely 2C) Mg-dependent beta isoform; Protein phosphatase type 1B (formely 2C), Mg-dependent, beta isoform; protein phosphatase, Mg2+/Mn2+ dependent, 1B; Ser/Thr protein phosphatase type 2C beta 2 isoform
Gene Aliases: PP2C-beta; PP2C-beta-X; Pp2c2; PP2CB; PP2CBETA; PPC2BETAX; PPM1B; Pppm1b
UniProt ID: (Human) O75688, (Rat) P35815
Entrez Gene ID: (Dog) 474573, (Human) 5495, (Rat) 24667
Molecular Function:
enzyme modulator
hydrolase
kinase inhibitor
kinase modulator
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.