Invitrogen
Manufacturer Code:PA587237
Catalog # PIPA587237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human PPM1A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IA; Mpp alpha; PP2C-alpha; Protein phosphatase 1A; protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform; Protein phosphatase 1A Protein phosphatase 2C isoform alpha PP2C-alpha Protein phosphatase IA PPM1A PPPM1A; protein phosphatase 1A, magnesium dependent, alpha isoform; Protein phosphatase 2C isoform alpha; Protein phosphatase IA; Protein phosphatase type 1A (formely 2C) Mg-dependent alpha isoform; Protein phosphatase type 1A (formely 2C), Mg-dependent, alpha isoform; protein phosphatase, Mg2+/Mn2+ dependent, 1A
Gene Aliases: 2310003C21Rik; 2900017D14Rik; AI427932; AU017636; MMPa-2; MPPa-1; PP2C-ALPHA; Pp2c1; PP2CA; PP2Calpha; PPM1A; PPPM1A
UniProt ID: (Human) P35813, (Mouse) P49443, (Rat) P20650
Entrez Gene ID: (Human) 5494, (Mouse) 19042, (Rat) 24666
Molecular Function: enzyme modulator hydrolase kinase inhibitor kinase modulator phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.