Invitrogen
Manufacturer Code:PA570085
Catalog # PIPA570085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human PPIP5K2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Diphosphoinositol pentakisphosphate kinase 2; HISPPD1 IP7K2 VIP2 VIP1 homolog 2 histidine acid phosphatase domain containing 1 histidine acid phosphatase domain-containing protein 1 hsVIP2 inositol heptaphosphate kinase 2 inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2 insP6 and PP-IP5 kinase; histidine acid phosphatase domain containing 1; Histidine acid phosphatase domain-containing protein 1; hsVIP2; inositol heptaphosphate kinase 2; Inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2; InsP6 and PP-IP5 kinase 2; mmVIP2; VIP1 homolog 2
Gene Aliases: AW555814; CFAP160; D330021B20; HISPPD1; IP7K2; KIAA0433; mKIAA0433; PPIP5K2; VIP2
UniProt ID: (Human) O43314, (Mouse) Q6ZQB6
Entrez Gene ID: (Rabbit) 100353531, (Dog) 488883, (Bovine) 780855, (Horse) 100064726, (Human) 23262, (Guinea pig) 100712709, (Rat) 501194, (Mouse) 227399
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.