Invitrogen
Manufacturer Code:PA529340
Catalog # PIPA529340
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 5 and 341 of Human HISPPD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Diphosphoinositol pentakisphosphate kinase 2; HISPPD1 IP7K2 VIP2 VIP1 homolog 2 histidine acid phosphatase domain containing 1 histidine acid phosphatase domain-containing protein 1 hsVIP2 inositol heptaphosphate kinase 2 inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2 insP6 and PP-IP5 kinase; Histidine acid phosphatase domain-containing protein 1; hsVIP2; inositol heptaphosphate kinase 2; Inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2; InsP6 and PP-IP5 kinase 2; VIP1 homolog 2
Gene Aliases: CFAP160; HISPPD1; IP7K2; KIAA0433; PPIP5K2; VIP2
UniProt ID: (Human) O43314
Entrez Gene ID: (Human) 23262
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.