OriGene
Manufacturer Code:TA505245
Catalog # 50-167-5765
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G3 |
Immunogen | Full length human recombit protein of human PPIL3 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cyclophilin J; cyclophilin-like protein 3; Cyclophilin-like protein PPIL3; CYPJ; Peptidyl-prolyl cis-trans isomerase-like 3; peptidylprolyl cis-trans isomerase-like protein 3; peptidylprolyl isomerase (cyclophilin)-like 3; peptidylprolyl isomerase -like 3; PPIase; PPIase-like protein 3; Rotamase PPIL3
Gene Aliases: CYPJ; PPIL3
UniProt ID: (Human) Q9H2H8
Entrez Gene ID: (Human) 53938
Molecular Function: isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.