Invitrogen
Manufacturer Code:PA530742
Catalog # PIPA530742
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 149 of Human PPIL3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cyclophilin J; cyclophilin-like 3; cyclophilin-like protein 3; Cyclophilin-like protein PPIL3; CYP10L; CyPJ; CYPJ PPIase PPIase-like protein 3 cyclophilin J cyclophilin-like protein 3 cyclophilin-like protein PPIL3 peptidyl-prolyl cis-trans isomerase-like 3 peptidylprolyl cis-trans isomerase-like protein 3 rotamase PPIL3; Peptidyl-prolyl cis-trans isomerase-like 3; peptidylprolyl cis-trans isomerase-like protein 3; peptidylprolyl isomerase (cyclophilin)-like 3; peptidylprolyl isomerase -like 3; PPIase; PPIase-like protein 3; Rotamase PPIL3
Gene Aliases: 2310076N22Rik; 2510026K04Rik; Cyp10l; CYPJ; PPIL3
UniProt ID: (Human) Q9H2H8, (Mouse) Q9D6L8
Entrez Gene ID: (Human) 53938, (Mouse) 70225
Molecular Function: isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.