Novus Biologicals
Manufacturer Code:NBP273572IR
Catalog # NBP273572IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C6 |
Immunogen | Full length recombinant protein of human PPIH (NP_006338) produced in E.coli. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: cyclophilin H; cyclophilin H CYP20 CypH CYPHpeptidyl prolyl isomerase H (cyclophilin H) EC 5.2.1.8 MGC5016 peptidyl-prolyl cis-trans isomerase H peptidylprolyl isomerase H (cyclophilin H) PPIase H Rotamase H Small nuclear ribonucleoprotein particle-specific cyclophilin H SnuCyp-20 USA-CyP SnuCyp-20 USA-CYPCYP-20 U-snRNP-associated cyclophilin SnuCyp-20 U-snRNP-associated cyclophilin SunCyp-20; CypH; Peptidyl-prolyl cis-trans isomerase H; peptidylprolyl isomerase H (cyclophilin H); PPIase H; Rotamase H; Small nuclear ribonucleoprotein particle-specific cyclophilin H; U-snRNP-associated cyclophilin SnuCyp-20; U-snRNP-associated cyclophilin SunCyp-20; USA-CYP; USA-CyP SnuCyp-20
Gene Aliases: CYP-20; CYP20; CYPH; PPIH; SnuCyp-20; USA-CYP
UniProt ID: (Human) O43447
Entrez Gene ID: (Human) 10465
Molecular Function: isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.