Proteintech
Manufacturer Code:116511AP150UL
Catalog # 50-172-7345
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PPIH Fusion Protein Ag2253 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cyclophilin H; CYP 20 CYP20 CYPH PPIase H PPIH Rotamase H SnuCyp 20 USA CYP; CypH; Peptidyl-prolyl cis-trans isomerase H; peptidylprolyl isomerase H (cyclophilin H); PPIase H; Rotamase H; Small nuclear ribonucleoprotein particle-specific cyclophilin H; U-snRNP-associated cyclophilin SnuCyp-20; U-snRNP-associated cyclophilin SunCyp-20; USA-CYP; USA-CyP SnuCyp-20
Gene Aliases: 1100001J08Rik; 2010111B15Rik; 4833408F11Rik; AI464484; CYP-20; CYP20; CYPH; D4Wsu43e; PPIH; RGD1564921; SnuCyp-20; USA-CYP
UniProt ID: (Human) O43447, (Mouse) Q9D868
Entrez Gene ID: (Human) 10465, (Rat) 366461, (Mouse) 66101
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.