Invitrogen
Manufacturer Code:PA545731
Catalog # PIPA545731
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human PPIG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CARS-cyclophilin; CARS-Cyp; CASP10; Clk-associating RS-cyclophilin; Cyclophilin G; Matrin cyclophilin; matrin cyclophilin (matrin-cyp); Matrin-cyp; Peptidyl-prolyl cis-trans isomerase G; peptidyl-prolyl isomerase G; peptidyl-prolyl isomerase G (cyclophilin G); peptidylprolyl isomerase G; peptidylprolyl isomerase G (cyclophilin G); Peptidylprolyl isomerase G (cyclophilin G) CARS-Cyp CYP MGC133241 SRCyp SCAF10; PPIase G; Rotamase G; SR-cyclophilin; SR-cyp; SR-related CTD-associated factor 10
Gene Aliases: AU019516; AU022200; B230312B02Rik; CARS-Cyp; CYP; PPIG; SCAF10; SRCyp
UniProt ID: (Human) Q13427, (Rat) O55035, (Mouse) A2AR02
Entrez Gene ID: (Dog) 607519, (Bovine) 783720, (Rabbit) 100341939, (Horse) 100052595, (Human) 9360, (Guinea pig) 100735043, (Rat) 83624, (Mouse) 228005
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.