Invitrogen
Manufacturer Code:PA5105260
Catalog # PIPA5105260
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PPIG corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU019516 AU022200 B230312B02Rik CARS-cyclophilin CARS-Cyp CASP10 Clk-associating RS-cyclophilin Cyclophilin G CYP Matrin cyclophilin matrin cyclophilin (matrin-cyp) Matrin-cyp MGC133241 OTTHUMP00000204761 OTTHUMP00000204762 peptidyl-prolyl cis-trans isomerase G peptidylprolyl isomerase G peptidyl-prolyl isomerase G peptidylprolyl isomerase G (cyclophilin G) peptidyl-prolyl isomerase G (cyclophilin G) PPIase G Ppig rotamase G SCAF10 SR-cyclophilin SRCyp SR-cyp SR-related CTD-associated factor 10; CARS-cyclophilin; CARS-Cyp; CASP10; Clk-associating RS-cyclophilin; Cyclophilin G; Matrin cyclophilin; matrin cyclophilin (matrin-cyp); Matrin-cyp; Peptidyl-prolyl cis-trans isomerase G; peptidyl-prolyl isomerase G; peptidyl-prolyl isomerase G (cyclophilin G); peptidylprolyl isomerase G; peptidylprolyl isomerase G (cyclophilin G); PPIase G; Rotamase G; SR-cyclophilin; SR-cyp; SR-related CTD-associated factor 10
Gene Aliases: AU019516; AU022200; B230312B02Rik; CARS-Cyp; CYP; PPIG; SCAF10; SRCyp
UniProt ID: (Human) Q13427, (Rat) O55035, (Mouse) A2AR02
Entrez Gene ID: (Human) 9360, (Rat) 83624, (Mouse) 228005
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.