Novus Biologicals
Manufacturer Code:NBP298654UV
Catalog # NB105008
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human PPIG fragment. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CARS-cyclophilin; CARS-cyclophilin CARS-CypPPIase G CASP10 Clk-associating RS-cyclophilin Cyclophilin G CYP EC 5.2.1.8 MGC133241 peptidyl-prolyl cis-trans isomerase G Peptidyl-prolyl isomerase G peptidylprolyl isomerase G (cyclophilin G) peptidyl-prolyl isomerase G (cyclophilin G) Rotamase G SCAF10 SR-cyclophilin SRcyp SR-cyp SR-related CTD-associated factor 10; CARS-Cyp; CASP10; Clk-associating RS-cyclophilin; Cyclophilin G; Peptidyl-prolyl cis-trans isomerase G; peptidyl-prolyl isomerase G (cyclophilin G); peptidylprolyl isomerase G (cyclophilin G); PPIase G; Rotamase G; SR-cyclophilin; SR-cyp; SR-related CTD-associated factor 10
Gene Aliases: CARS-Cyp; CYP; PPIG; SCAF10; SRCyp
UniProt ID: (Human) Q13427
Entrez Gene ID: (Human) 9360
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.