OriGene
Manufacturer Code:CF809029
Catalog # 50-166-9232
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C4 |
Immunogen | Full length human recombit protein of human PPIF produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cyclophilin 3; Cyclophilin D; Cyclophilin F; CyP-D; CyP-M; CYP3 CyP-M Cyp-D CypD; Mitochondrial cyclophilin; Peptidyl-prolyl cis-trans isomerase F, mitochondrial; peptidyl-prolyl cis-trans isomerase, mitochondrial; PPIase F; Rotamase F
Gene Aliases: Cyp-D; CyP-M; CYP3; CypD; PPIF
UniProt ID: (Human) P30405
Entrez Gene ID: (Human) 10105
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.