Invitrogen
Manufacturer Code:PA127785
Catalog # PIPA127785
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to human Peptidyl-prolyl cis-trans isomerase. mitochondrial. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C,Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: cyclophilin 3; Cyclophilin D; Cyclophilin F; CyP-D; CyP-M; Mitochondrial cyclophilin; Peptidyl-prolyl cis-trans isomerase F, mitochondrial; peptidyl-prolyl cis-trans isomerase, mitochondrial; PPIase F; Rotamase F; RP11-342M3.4 CYP3 Cyp-D FLJ90798 MGC117207
Gene Aliases: Cyp-D; CyP-M; CYP3; CypD; PPIF
UniProt ID: (Human) P30405
Entrez Gene ID: (Human) 10105
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.