Invitrogen
Manufacturer Code:PA522375
Catalog # PIPA522375
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 18 and 277 of Liprin alpha 1 (Uniprot IDNo.Q13136) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LAR-interacting protein 1; LIP-1; LIP.1 LIP1 LIPRIN LAR-interacting protein 1 LIP-1 Liprin-alpha1 PTPRF-interacting protein alpha-1 liprin-alpha-1 protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-1; liprin; Liprin-alpha-1; Liprin-alpha1; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-1; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 1; PTPRF-interacting protein alpha-1
Gene Aliases: C030014K08Rik; C87158; LIP.1; LIP1; LIPRIN; PPFIA1
UniProt ID: (Human) Q13136
Entrez Gene ID: (Human) 8500, (Mouse) 233977
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.