Invitrogen
Manufacturer Code:PA127622
Catalog # PIPA127622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to human Liprin-alpha-1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C,Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ41337 FLJ42630 FLJ43474 LIP.1 LIP1 LIPRIN MGC26800; LAR-interacting protein 1; LIP-1; Liprin-alpha-1; Liprin-alpha1; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-1; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 1; PTPRF-interacting protein alpha-1
Gene Aliases: LIP.1; LIP1; LIPRIN; PPFIA1
UniProt ID: (Human) Q13136
Entrez Gene ID: (Human) 8500
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.