Invitrogen
Manufacturer Code:PA598076
Catalog # PIPA598076
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Serine/threonine-protein phosphatase with EF-hands 1 protein (301-500AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DRES10; PP7; PPEF; PPEF 1; PPEF-1; protein phosphatase 7, catalytic subunit, alpha isozyme; Protein phosphatase with EF calcium-binding domain; protein phosphatase, EF-hand calcium binding domain 1; protein phosphatase, serine/threonine type, with EF-hands; serine/threonine protein phosphatase 7; Serine/threonine-protein phosphatase 7; Serine/threonine-protein phosphatase with EF-hands 1
Gene Aliases: Dres10; PP7; PPEF; PPEF-1; PPEF1; PPP7C; PPP7CA
UniProt ID: (Human) O14829, (Mouse) O35655
Entrez Gene ID: (Human) 5475, (Mouse) 237178
Molecular Function: calcium-binding protein hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.