Novus Biologicals
Manufacturer Code:NBP219897
Catalog # NBP219897
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human PPEF1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.3.16 PP7 PPEF PPEF-1 PPP7C PPP7CA protein phosphatase 7 catalytic subunit alpha isozyme Protein phosphatase with EF calcium-binding domain protein phosphatase EF-hand calcium binding domain 1 protein phosphatase serine/threonine type with EF-hands serine/threonine protein phosphatase 7 Serine/threonine-protein phosphatase 7 serine/threonine-protein phosphatase with EF-hands 1; PP7; PPEF; PPEF-1; protein phosphatase 7, catalytic subunit, alpha isozyme; Protein phosphatase with EF calcium-binding domain; protein phosphatase, EF-hand calcium binding domain 1; protein phosphatase, serine/threonine type, with EF-hands; serine/threonine protein phosphatase 7; Serine/threonine-protein phosphatase 7; Serine/threonine-protein phosphatase with EF-hands 1
Gene Aliases: PP7; PPEF; PPEF1; PPP7C; PPP7CA
UniProt ID: (Human) O14829
Entrez Gene ID: (Human) 5475
Molecular Function: calcium-binding protein hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.