Invitrogen
Manufacturer Code:MA536234
Catalog # PIMA536234
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7G13 |
Immunogen | A synthetic peptide corresponding to a sequence of human PPCS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 6330579B17Rik AI413471 AI790434 Coab Phosphopantothenate--cysteine ligase Phosphopantothenoylcysteine synthetase PPC synthetase PPCS RGD1311399; Phosphopantothenate--cysteine ligase; Phosphopantothenoylcysteine synthetase; PPC synthetase
Gene Aliases: 6330579B17Rik; AI413471; AI790434; COAB; PPCS; RGD1311399
UniProt ID: (Human) Q9HAB8, (Mouse) Q8VDG5
Entrez Gene ID: (Human) 79717, (Mouse) 106564, (Rat) 298490
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.