Invitrogen
Manufacturer Code:PA542852
Catalog # PIPA542852
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PPCDC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CoaC; Phosphopantothenoylcysteine decarboxylase; PPC-DC
Gene Aliases: 1810057I13Rik; 8430432M10Rik; COAC; MDS018; PPC-DC; PPCDC; RGD1306267; UNQ9365/PRO34154
UniProt ID: (Human) Q96CD2, (Mouse) Q8BZB2
Entrez Gene ID: (Horse) 100051247, (Bovine) 614957, (Dog) 487653, (Rabbit) 100343612, (Human) 60490, (Rat) 363069, (Guinea pig) 100712891, (Mouse) 66812
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.