Novus Biologicals
Manufacturer Code:NBP15648920UL
Catalog # NBP15648920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to PPCDC(phosphopantothenoylcysteine decarboxylase) The peptide sequence was selected from the N terminal of PPCDC. Peptide sequence VTTERAKHFYSPQDIPVTLYSDADEWEIWKSRSDPVLHIDLRRWADLLLV. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CoaC; coaC EC 4.1.1.36 FLJ14585 MDS018 phosphopantothenoylcysteine decarboxylase PPC-DC; Phosphopantothenoylcysteine decarboxylase; PPC-DC
Gene Aliases: COAC; MDS018; PPC-DC; PPCDC; UNQ9365/PRO34154
UniProt ID: (Human) Q96CD2
Entrez Gene ID: (Human) 60490
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.