Novus Biologicals
Manufacturer Code:NBP300333AF532
Catalog # NB065598
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant PPCDC (AAH14409.1 Met1-Ser204). |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CoaC; coaC EC 4.1.1.36 FLJ14585 MDS018 phosphopantothenoylcysteine decarboxylase PPC-DC; Phosphopantothenoylcysteine decarboxylase; PPC-DC
Gene Aliases: COAC; MDS018; PPC-DC; PPCDC; UNQ9365/PRO34154
UniProt ID: (Human) Q96CD2
Entrez Gene ID: (Human) 60490
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.