Invitrogen
Manufacturer Code:PA552549
Catalog # PIPA552549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PPBP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-TG; Beta-thromboglobulin; C-X-C motif chemokine 7; chemokine (C-X-C motif) ligand 7; Connective tissue-activating peptide III; Connective tissue-activating peptide III(1-81); CTAP-III; CTAP-III(1-81); CXC chemokine ligand 7; LA-PF4; LDGF; Leukocyte-derived growth factor; Low-affinity platelet factor IV; Macrophage-derived growth factor; MDGF; NAP-2; NAP-2(1-63); NAP-2(1-66); NAP-2(73); NAP-2(74); Neutrophil-activating peptide 2; Neutrophil-activating peptide 2(1-63); Neutrophil-activating peptide 2(1-66); Neutrophil-activating peptide 2(73); Neutrophil-activating peptide 2(74); neutrophil-activating peptide-2; PBP; Platelet basic protein; Platelet basic protein PBP C-X-C motif chemokine 7 Leukocyte-derived growth factor LDGF Macrophage-derived growth factor MDGF Small-inducible cytokine B7 Connective tissue-activating peptide III CTAP-III LA-PF4 Low-affinity platelet factor IV TC-2 Connective tissue-activating peptide III(1-81) CTAP-III(1-81) PPBP CTAP3 CXCL7 SCYB7 TGB1 THBGB1; pro-platelet basic protein (chemokine (C-X-C motif) ligand 7); small inducible cytokine B7; small inducible cytokine subfamily B, member 7; Small-inducible cytokine B7; TC-1; TC-2; thrombocidin 1; thrombocidin 2; thromboglobulin, beta-1
Gene Aliases: B-TG1; Beta-TG; CTAP-III; CTAP3; CTAPIII; CXCL7; LA-PF4; LDGF; MDGF; NAP-2; PBP; PPBP; SCYB7; TC1; TC2; TGB; TGB1; THBGB; THBGB1
UniProt ID: (Human) P02775
Entrez Gene ID: (Human) 5473
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.