Invitrogen
Manufacturer Code:MA525978
Catalog # PIMA525978
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B8 |
Immunogen | Human recombit protein fragment corresponding to amino acids 42-278 of PPAT produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Amidophosphoribosyltransferase; amidophosphoribosyltransferase proprotein; ATase; ATASE GPAT PRAT amidophosphoribosyltransferase glutamine PRPP amidotransferase glutamine phosphoribosylpyrophosphatate amidotransferase glutamine phosphoribosylpyrophosphate amidotransferase; glutamine phosphoribosylpyrophosphatate amidotransferase; Glutamine phosphoribosylpyrophosphate amidotransferase; glutamine PRPP amidotransferase; GPAT
Gene Aliases: 5730454C12Rik; AA408689; AA675351; AI507113; ATASE; C79945; EGK_15868; GPAT; PPAT; PRAT
UniProt ID: (Human) Q06203, (Rat) P35433
Entrez Gene ID: (Human) 5471, (Dog) 611407, (Rhesus monkey) 694868, (Rat) 117544, (Mouse) 231327
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.