Invitrogen
Manufacturer Code:PA585594
Catalog # PIPA585594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human PGC1 beta. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: peroxisome proliferative activated receptor, gamma, coactivator 1 beta; Peroxisome proliferator-activated receptor gamma coactivator 1-beta; peroxisome proliferator-activated receptor gamma coactivator 1beta-2a; PGC-1-beta; PPAR-gamma coactivator 1-beta; PPARG coactivator 1 beta Antibody PGC1(beta) Antibody ERRL1 Antibody PGC1B Antibody PERC Antibody PPARGC1B Antibody; PPARGC-1-beta
Gene Aliases: Perc; Pgc1; Pgc1b; PGC1beta; Ppargc1; Ppargc1b
UniProt ID: (Rat) Q811R2
Entrez Gene ID: (Rat) 291567
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.