Abnova
Manufacturer Code:H00010891M17
Catalog # 89-002-783
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2G8 |
Immunogen | PPARGC1A (NP_037393 689 a.a. ~ 798 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant PPARGC1A.
For Research Use Only
Protein Aliases: L-PGC-1alpha; LEM6; Ligand effect modulator 6; ligand effect modulator-6; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-3ext; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-8a; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5-NT; Peroxisome proliferator-activated receptor gamma coactivator 1-alpha; peroxisome proliferator-activated receptor gamma, coactivator 1 alpha; PGC-1(alpha); PGC-1-alpha; PGC-1v; PGC1; PGC1A; PPAR gamma coactivator variant form; PPARgamma coactivator 1alpha; PPARGC-1-alpha; PPARGC1
Gene Aliases: LEM6; PGC-1(alpha); PGC-1alpha; PGC-1v; PGC1; PGC1A; PPARGC1; PPARGC1A
UniProt ID: (Human) Q3LIG1
Entrez Gene ID: (Human) 10891
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.