Abnova
Manufacturer Code:MAB12607
Catalog # 89-934-794
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A4A9 + 1E6A1 |
Immunogen | Recombinant protein corresponding to amino acids 170-270 of human PPARG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against human PPARG.
For Research Use Only
Protein Aliases: CIMT1; NR1C3; Nuclear receptor subfamily 1 group C member 3; peroxisome proliferator-activated nuclear receptor gamma variant 1; Peroxisome proliferator-activated receptor gamma; PPAR-gamma; PPARG1; PPARG2; PPARgamma
Gene Aliases: CIMT1; GLM1; NR1C3; PPARG; PPARG1; PPARG2; PPARgamma
UniProt ID: (Human) P37231
Entrez Gene ID: (Human) 5468
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.