Proteintech
Manufacturer Code:601931IG150UL
Catalog # 50-173-6296
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B10E1 |
Immunogen | PPARD Fusion Protein Ag16770 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FAAR NR1C2 NUC1 NUCI NUCII Nuclear hormone receptor 1 PPAR beta PPAR delta PPARB PPARD; NUC1; NUCI; Nuclear hormone receptor 1; Nuclear receptor subfamily 1 group C member 2; peroxisome proliferator-activated nuclear receptor beta/delta variant 2; Peroxisome proliferator-activated receptor beta; Peroxisome proliferator-activated receptor delta; PPAR-beta; PPAR-delta; PPARdelta/beta
Gene Aliases: FAAR; NR1C2; NUC-1; NUC1; NUCI; NUCII; PPAR-beta; PPAR-delta; PPAR[b]; PPARB; Pparb/d; PPARD; PPARdelta
UniProt ID: (Human) Q03181, (Mouse) P35396
Entrez Gene ID: (Human) 5467, (Mouse) 19015
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.