Abnova
Manufacturer Code:PAB11321
Catalog # 89-115-014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 1-18 of mouse Ppara. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Ppara.
For Research Use Only
Protein Aliases: 4933429D07Rik; AW742785; Nr1c1; Nuclear receptor subfamily 1 group C member 1; Peroxisome proliferator-activated receptor alpha; Ppar; PPAR-alpha; PPARalpha
Gene Aliases: 4933429D07Rik; AW742785; Nr1c1; Ppar; PPAR-alpha; Ppara; PPARalpha
UniProt ID: (Mouse) P23204
Entrez Gene ID: (Mouse) 19013
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.