Invitrogen
Manufacturer Code:MA535338
Catalog # PIMA535338
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0155 |
Immunogen | A synthesized peptide derived from human PPAR gamma. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CIMT1 GLM1 HUMPPARG Nr1c3 Nuclear receptor subfamily 1 group C member 3 OTTHUMP00000185032 OTTHUMP00000185036 Peroxisome proliferator-activated receptor gamma PPAR gamma Pparg PPARG1 PPARG2 PPARgamma PPAR-gamma PPARgamma2 PPAR-gamma2 PPARy; Nuclear receptor subfamily 1 group C member 3; peroxisome proliferator activated receptor gamma; peroxisome proliferator activator receptor, gamma; Peroxisome proliferator-activated receptor gamma; PPAR-gamma
Gene Aliases: Nr1c3; Pparg
UniProt ID: (Rat) O88275
Entrez Gene ID: (Rat) 25664
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.