Invitrogen
Manufacturer Code:PA525757
Catalog # PIPA525757
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PPARG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CIMT1 GLM1 NR1C3 PPARG1 PPARG2 PPARgamma PPAR gamma PPAR-gamma nuclear receptor subfamily 1 group C member 3 peroxisome proliferator-activated nuclear receptor gamma variant 1 peroxisome proliferator-activated receptor gamma 1; Nuclear receptor subfamily 1 group C member 3; peroxisome proliferator activated receptor gamma 2; peroxisome proliferator activated receptor gamma 4; peroxisome proliferator-activated nuclear receptor gamma variant 1; Peroxisome proliferator-activated receptor gamma; PPAR-gamma
Gene Aliases: CIMT1; GLM1; NR1C3; PPAR-gamma; PPAR-gamma2; PPARG; PPARG1; PPARG2; PPARgamma; PPARgamma2
UniProt ID: (Human) P37231, (Mouse) P37238
Entrez Gene ID: (Human) 5468, (Mouse) 19016
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.