Invitrogen
Manufacturer Code:PA19576
Catalog # 01-674-452
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide based on human PPARg protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: HUMPPARG NR1C3 PPARG1 PPARG2; Nuclear receptor subfamily 1 group C member 3; peroxisome proliferator-activated nuclear receptor gamma variant 1; Peroxisome proliferator-activated receptor gamma; PPAR-gamma
Gene Aliases: CIMT1; GLM1; NR1C3; PPARG; PPARG1; PPARG2; PPARgamma
UniProt ID: (Human) P37231
Entrez Gene ID: (Human) 5468
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.