Novus Biologicals
Manufacturer Code:NBP30321420UL
Catalog # NB093577
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 102-361 of human PPAR delta/NR1C2 (NP_803184.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FAAR MGC3931 NR1C2PPAR-delta NUC1PPAR-beta NUCI NUCII Nuclear hormone receptor 1 Nuclear receptor subfamily 1 group C member 2 peroxisome proliferative activated receptor delta Peroxisome proliferator-activated receptor beta peroxisome proliferator-activated receptor delta PPARB; NUC1; NUCI; Nuclear hormone receptor 1; Nuclear receptor subfamily 1 group C member 2; peroxisome proliferator-activated nuclear receptor beta/delta variant 2; Peroxisome proliferator-activated receptor beta; Peroxisome proliferator-activated receptor delta; PPAR-beta; PPAR-delta
Gene Aliases: FAAR; NR1C2; NUC1; NUCI; NUCII; PPARB; PPARD
UniProt ID: (Human) Q03181
Entrez Gene ID: (Human) 5467
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.