Invitrogen
Manufacturer Code:PA518299
Catalog # PIPA518299
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (CHPLLQEIYKDMY) corresponding to the C-terminus amino acids of PPARD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: NUC1; NUCI; Nuclear hormone receptor 1; Nuclear receptor subfamily 1 group C member 2; peroxisome proliferator activated receptor delta; peroxisome proliferator activator receptor beta; peroxisome proliferator activator receptor, delta; peroxisome proliferator-activated nuclear receptor beta/delta variant 2; Peroxisome proliferator-activated receptor beta; Peroxisome proliferator-activated receptor delta; PPAR-beta; PPAR-delta; PPARD FAAR NUC1 NUCI NR1C2 NUCII PPARB MGC3931 PPAR-beta peroxisome proliferative activated receptor delta nuclear hormone receptor 1 peroxisome proliferative activated receptor delta OTTHUMP00000016256 OTTHUMP00000016257
Gene Aliases: FAAR; NR1C2; NUC1; NUCI; NUCII; PPARB; PPARD
UniProt ID: (Human) Q03181
Entrez Gene ID: (Human) 5467, (Rat) 25682
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.