Novus Biologicals
Manufacturer Code:NBP273567H
Catalog # NBP273567H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E8 |
Immunogen | Recombinant protein fragment corresponding to amino acids 145-420 of human PPAR alpha/NR1C1 (NP_005027) produced in E.coli. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: alpha hPPAR NR1C1MGC2237 Nuclear receptor subfamily 1 group C member 1 peroxisome proliferator-activated receptor alpha PPARalpha PPAR-alpha PPARMGC2452; Nuclear receptor subfamily 1 group C member 1; peroxisome proliferative activated receptor, alpha; peroxisome proliferator-activated nuclear receptor alpha variant 3; Peroxisome proliferator-activated receptor alpha; PPAR-alpha
Gene Aliases: hPPAR; NR1C1; PPAR; PPARA; PPARalpha
UniProt ID: (Human) Q07869
Entrez Gene ID: (Human) 5465
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.