Novus Biologicals
Manufacturer Code:NB300537AF350
Catalog # NB30537A350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3B6/PPAR |
Immunogen | Purified recombinant PPAR alpha protein. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: alpha hPPAR NR1C1MGC2237 Nuclear receptor subfamily 1 group C member 1 peroxisome proliferator-activated receptor alpha PPARalpha PPAR-alpha PPARMGC2452; Nuclear receptor subfamily 1 group C member 1; peroxisome proliferative activated receptor, alpha; peroxisome proliferator-activated nuclear receptor alpha variant 3; Peroxisome proliferator-activated receptor alpha; PPAR-alpha
Gene Aliases: hPPAR; NR1C1; PPAR; PPARA; PPARalpha
UniProt ID: (Human) Q07869
Entrez Gene ID: (Human) 5465
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.